Order Entry
Germany
Orders LinkContactUsLinkComponent
Ampicillin

Ampicillin

Glass bottle for solids, 10 g
Catalog # SERA13397.01
Marketsource
Supplier:   Serva
Ampicillin
Catalog # SERA13397.01
Supplier:  Serva
Marketsource
CAS Number:  69-53-4
Ampicillin

Glass bottle for solids, 10 g

CAS Number:  69-53-4
Price
67,60 €
Log in for availability
1 * 10 g
Product Details & Documents
  • CAS Number:69-53-4
  • Formula:C₁₆H₁₉N₃O₄S
  • MDL Number:MFCD00005175
  • Molecular Weight:349.4 g/mol
  • EINECS:200-709-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.