Agar
5 kg
Catalog # SERA11396.04
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: Agar Agar
- CAS Number:9002-18-0
- MDL Number:MFCD00081288
- Merck Index:13,00184
- Melting Pt:90 °C
- Storage Temperature:Ambient
- EINECS:232-658-1
Specifications
Specifications
- Pk:5 kg



