Order Entry
Germany
Orders LinkContactUsLinkComponent
Agar

Agar

Glass bottle for solids, 250 g
Catalog # SERA11393.03
Marketsource
Supplier:   Serva
Agar
Catalog # SERA11393.03
Supplier:  Serva
Marketsource
CAS Number:  9002-18-0
Agar
Agar

Glass bottle for solids, 250 g

CAS Number:  9002-18-0
Price
190,00 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.