Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Nalgene™ Screw Caps

Replacement closure, 53 mm, Clear, PP
  NALG71-2174-0530
Nalgene™ Screw Caps
  NALG71-2174-0530
Nalgene™ Screw Caps
Nalgene™ Screw Caps

Replacement closure, 53 mm, Clear, PP

 
53,20 €
   
1 * 2 ST
 

PP, coloured.

  • Very good chemical resistance
  • Heat resistant up to 135 °C
  • Autoclavable

Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.

 
  • Cap Size:
    53 mm
  • Colour:
    Clear
  • Description:
    Replacement closure
  • Material:
    PP
  • Seq:
    49