VWR®, Calibration Weight Sets, Class E2
1 mg - 1 kg, 25, 2111,11 g
611-8747
Weights made from solid, anti-magnetic, polished stainless steel. Suitable for high resolution electronic balances.
- Wire weights up to 500 mg in cobalt alloy
- Identification number guarantees traceability for weights from 1 g upwards
- Laser engraving improves legibility of the markings
- Available with or without COFRAC certificate
Conform to OILM R111 recommendations.
: Conform to OILM R111 recommendations.
: All weight sets listed in the 'set contents' table are available with or without COFRAC certificate - please visit website or contact your local Avantor sales office. Weights are also available individually - please search for product 'Calibration weights, Class E2'.
- Weight set:1 mg - 1 kg
- Seq:06
- No. of weights:25
- Total weight:2111,11 g



