VWR®, Block Weights, Class M1
5 kg, 250 ± mg, With certificate
611-8324
Rectangular weights made from cast iron (density 7,2 g/cm³).
- Indelible unique identification number guarantees traceability
Colour: Grey.
Conforms to OILM, COFRAC accredited
: Conforms to OIML, COFRAC accredited.
: Block weights are supplied with cardboard packaging to protect the weights during transportation.
- Weight:5 kg
- Seq:005
- Tolerance:250 ± mg
- Type:With certificate



