Order Entry
Germany
Orders LinkContactUsLinkComponent
Dropping bottles, LDPE, Nalgene®

Dropping bottles, LDPE, Nalgene®

30 ml
Catalog # 215-7872
   Low Manufacturing Impact
Dropping bottles, LDPE, Nalgene®
Catalog # 215-7872
   Low Manufacturing Impact
Dropping bottles, LDPE, Nalgene®
Dropping bottles, LDPE, Nalgene®

30 ml

Price
58,80 €
Log in for availability
1 * 12 ST
Product Details & Documents

LDPE, translucent, with dropping seal and PP cap.

  • Good chemical resistance, heat resistant up to 80 °C
  • Tethered nozzle cannot be lost
  • Leakproof

Only dispenses single drops. Easy filling.

Specifications
  • Capacity:
    30 ml
  • Cat. No.:
    215-7872
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.