3-Fluoropyrocatechol
Glass bottle for solids, 10 g
Catalog # MAYBSB01505.0010
Marketsource
Supplier: Thermo Scientific
3-Fluoropyrocatechol
Catalog # MAYBSB01505.0010
Supplier: Thermo Scientific
Marketsource
CAS Number: 363-52-0
Jump to:
Product Details & Documents
Synonyms: 3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene
- CAS Number:363-52-0
- Formula:C₆H₅FO₂
- MDL Number:MFCD00042582
- Molecular Weight:128.1 g/mol
- Boiling Pt:218 °C (1013 hPa)
- Melting Pt:71 °C
- Density:1,514 g/cm³ (20 °C)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



