Order Entry
Germany
Orders LinkContactUsLinkComponent
Büchner funnels, DURAN®

Büchner funnels, DURAN®

48 mm, 70 ml, 45 mm
Catalog # 511-0335
Supplier:   DWK Life Sciences
Büchner funnels, DURAN®
Catalog # 511-0335
Supplier:  DWK Life Sciences
Büchner funnels, DURAN®
Büchner funnels, DURAN®

48 mm, 70 ml, 45 mm

Price
96,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass 3.3, clear.

For filter papers or membrane filtration discs.

Specifications
  • Plate Ø:
    48 mm
  • Capacity:
    70 ml
  • For filter Ø:
    45 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.