Rotary evaporator receiving flasks
500 ml, KS 35/20
Catalog # 531-9144
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, clear or plastic-coated.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Spherical joint KS 35/20, centred, suitable for all rotary evaporators.
Documents
BSE TSE Glass Columns
BSE/TSE-Zertifikate
Specifications
- Socket:KS 35/20
- Capacity:500 ml
- Seq:03