Spherical ground joints
120 mm, S 19, -
Catalog # 201-4223
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, clear.
- Excellent sealing properties
- Minimal requirement for joint lubrication
- Ideal for use as flexible connectors
Spherical cup and ball joint with inch markings. Fused edges. Excellent chemical resistance. High temperature resistance. Minimal thermal expansion giving relatively high resistance to temperature changes.
DIN 12244 (201-4215 and 201-0128 in addition to DIN), ISO 641
: DIN 12244 (201-4215 and 201-0128 in addition to DIN), ISO 641.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Ball:S 19
- Socket:-
- Total length:120 mm