Order Entry
Germany
Orders LinkContactUsLinkComponent
Sodium hydroxide 30% in aqueous solution

Sodium hydroxide 30% in aqueous solution

Plastic bottle, 5 L
Catalog # KRAF05488.5020
Corrosive
Supplier:   Bernd Kraft
Sodium hydroxide 30% in aqueous solution
Catalog # KRAF05488.5020
Supplier:  Bernd Kraft
CAS Number:  1310-73-2
Sodium hydroxide 30% in aqueous solution

Plastic bottle, 5 L

CAS Number:  1310-73-2
Price
Inquire For Price
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:119…135 °C (1013 hPa)
  • Melting Pt:9…10 °C
  • Density:1,53 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.