Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer solutions, ready to use

Buffer solutions, ready to use

9,0, Borate, Colourless, 20 °C, Plastic bottle, 10 L
Catalog # KRAF03088.6400
Supplier:   Bernd Kraft
Buffer solutions, ready to use
Catalog # KRAF03088.6400
Supplier:  Bernd Kraft
CAS Number:  
Buffer solutions, ready to use
Buffer solutions, ready to use

9,0, Borate, Colourless, 20 °C, Plastic bottle, 10 L

CAS Number:  
Price
188,00 €
Log in for availability
1 * 10 L
Product Details & Documents
  • NIST traceable

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Borate
  • Pk:
    10 L
  • Reference temperature:
    20 °C
  • Colour:
    Colourless
  • pH value:
    9,0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.