Order Entry
Germany
Orders LinkContactUsLinkComponent
Stirring rods

Stirring rods

Stirring rods, PVC, PVC, 7 mm, 300 mm
Catalog # 442-4151
Supplier:   KARTELL
Stirring rods
Catalog # 442-4151
Supplier:  KARTELL
Stirring rods
Stirring rods

Stirring rods, PVC, PVC, 7 mm, 300 mm

Price
9,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

PVC.

  • Rigid and unbreakable
  • Ideal replacement for glass rods
Specifications
  • Ø:
    7 mm
  • Description:
    Stirring rods, PVC
  • Length:
    300 mm
  • Material:
    PVC
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.