Stirring rods
Stirring rods, PVC, PVC, 7 mm, 250 mm
442-4150
: KARTELL
PVC.
- Rigid and unbreakable
- Ideal replacement for glass rods
- Ø:7 mm
- Description:Stirring rods, PVC
- Length:250 mm
- Material:PVC
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



