Order Entry
Germany
Orders LinkContactUsLinkComponent
Tanks

Tanks

Grey, 27 l, 340×455×250 mm
Catalog # 216-0256
Supplier:   KARTELL
Tanks
Catalog # 216-0256
Supplier:  KARTELL
Tanks
Tanks

Grey, 27 l, 340×455×250 mm

Price
91,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Made of HDPE, grey.

  • Moulded grips
  • Sturdy

Useful general purpose tanks that can be stacked for storage.

Specifications
  • Capacity:
    27 l
  • Colour:
    Grey
  • W×D×H:
    340×455×250 mm