Orders LinkContactUsLinkComponent
Microplates, 96-well
Microplates, 96-well
96-well plates, V-bottom, aseptic, Clear, 288 µl, 96
Catalog # KART2622
Supplier:   KARTELL
Microplates, 96-well
Catalog # KART2622
Supplier:  KARTELL
Microplates, 96-well
Microplates, 96-well
96-well plates, V-bottom, aseptic, Clear, 288 µl, 96
Price
113,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
Multiple-well plates (96 sample wells) in clear PS. Available in three shapes: flat bottom, U bottom and V bottom wells.
  • Aseptic or sterile
  • Single-wrapped
Ordering information: PS covers are sold separately.
Specifications
  • No. of wells:
    96
  • Volume:
    288 µl
  • Colour:
    Clear
  • Description:
    96-well plates, V-bottom, aseptic
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Lids for 96 well microtitre plates, PS, aseptic
    Lids for 96 well microtitre plates, PS, aseptic
    KART2623
    Log in for availability
    1 * 50 ST - 93,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Lids for 96 well microtitre plates, PS, sterile
    Lids for 96 well microtitre plates, PS, sterile
    KART2623/S
    Log in for availability
    1 * 50 ST - 97,30 €