Quinoline 99%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:91-22-5
- Formula:C₉H₇N
- MDL Number:MFCD00006736
- Molecular Weight:129.16 g/mol
- Merck Index:13,08160
- Boiling Pt:237 °C (760 mmHg)
- Melting Pt:–15 °C
- Density:1.095 g/cm³
- Flash Pt:101 °C
- Storage Temperature:Ambient
- EINECS:202-051-6
- UN:2656
- ADR:6.1,III
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless to dark brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6230 to 1.6270 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



