Sodium hydroxide 98.5%, pellets for analysis
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Sodium hydroxide 98.5%, pellets for analysis
Catalog # ACRO206060025
Supplier: Thermo Fisher Scientific
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Boiling Pt:1390 °C (760 mmHg)
- Melting Pt:318 °C
- Density:2,13 g/cm³ (20 °C)
- Storage Temperature:Ambient
- UN:1823
- ADR:8,II
Documents
ACRO20606
SDS
Specifications
Specification Test Results
- Appearance:White Pellets
- Titration with HCl:≥98.0 %
- Chloride (Cl):≤70 ppm
- Phosphate (PO4):≤3 ppm
- Sulfate (SO4):≤40 ppm
- Sodium carbonate (Na2CO3):≤0.5 %
- Aluminium (Al):≤5 ppm
- Calcium (Ca):≤10 ppm
- Iron (Fe):≤8 ppm
- Nickel (Ni):≤3 ppm
- Potassium (K):≤0.1 %
- Silver (Ag):≤1 ppm
- Heavy metals (as Pb):≤10 ppm
- Silicon dioxide (SiO2):≤20 ppm
- Nitrogen compounds (as N):≤0.0003 %
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



