Order Entry
Germany
Orders LinkContactUsLinkComponent
Sodium hydroxide 98.5%, pellets for analysis

Sodium hydroxide 98.5%, pellets for analysis

Plastic bottle, 1 kg
Catalog # ACRO206060010
Corrosive
Sodium hydroxide 98.5%, pellets for analysis
Catalog # ACRO206060010
CAS Number:  1310-73-2
Sodium hydroxide 98.5%, pellets for analysis
Sodium hydroxide 98.5%, pellets for analysis

Plastic bottle, 1 kg

CAS Number:  1310-73-2
Price
38,60 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:1390 °C (760 mmHg)
  • Melting Pt:318 °C
  • Density:2,13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • UN:1823
  • ADR:8,II

Documents

Specifications

Specification Test Results

  • Appearance:
    White Pellets
  • Titration with HCl:
    ≥98.0 %
  • Chloride (Cl):
    ≤70 ppm
  • Phosphate (PO4):
    ≤3 ppm
  • Sulfate (SO4):
    ≤40 ppm
  • Sodium carbonate (Na2CO3):
    ≤0.5 %
  • Aluminium (Al):
    ≤5 ppm
  • Calcium (Ca):
    ≤10 ppm
  • Iron (Fe):
    ≤8 ppm
  • Nickel (Ni):
    ≤3 ppm
  • Potassium (K):
    ≤0.1 %
  • Silver (Ag):
    ≤1 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Silicon dioxide (SiO2):
    ≤20 ppm
  • Nitrogen compounds (as N):
    ≤0.0003 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.