Potassium nitrate 99+% for analysis
Plastic bottle, 1 kg
Supplier: Thermo Fisher Scientific
Potassium nitrate 99+% for analysis
Catalog # ACRO205910010
Supplier: Thermo Fisher Scientific
CAS Number: 7757-79-1
Jump to:
Product Details & Documents
Synonyms: Nitrate of Potash
- CAS Number:7757-79-1
- Formula:KNO₃
- MDL Number:MFCD00011409
- Molecular Weight:101.1 g/mol
- Merck Index:13,07733
- Boiling Pt:400 °C (760 mmHg)
- Melting Pt:334 °C
- Density:2,109 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-818-8
- UN:1486
- ADR:5.1,III
- REACH:01-2119488224-35
Documents
ACRO20591
SDS
Specifications
Specification Test Results
- Appearance:White Crystals
- pH:5.5 to 8 (5 % at 20°C)
- Titration after Ion exchange:≥99.0 %
- Chloride (Cl):≤20 ppm
- Iodate (IO3):≤5 ppm
- Nitrite (NO2):≤10 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤10 ppm
- Sodium (Na):≤200 ppm
- Heavy metals (as Pb):≤10 ppm
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



