n-Eicosane 99%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: N-Eicosane
- CAS Number:112-95-8
- Formula:H₃C(CH₂)₁₈CH₃
- MDL Number:MFCD00009344
- Molecular Weight:282.55 g/mol
- Boiling Pt:220 °C (30 mmHg)
- Melting Pt:36…38 °C
- Density:0.7886 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
- EINECS:204-018-1
Documents
ACRO20437
SDS
Specifications
Specification Test Results
- Appearance:Transparent white Crystalline powder or flakes
- Infrared spectrum:Conforms
- Melting point:36 to 38 °C
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



