n-Eicosane 99%
Glass bottle for solids, 2 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: N-Eicosane
- CAS Number:112-95-8
- Formula:H₃C(CH₂)₁₈CH₃
- MDL Number:MFCD00009344
- Molecular Weight:282.55 g/mol
- Boiling Pt:220 °C (30 mmHg)
- Melting Pt:36…38 °C
- Density:0.7886 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
- EINECS:204-018-1
Documents
ACRO20437
SDS
Specifications
Specification Test Results
- Appearance:Transparent white Crystalline powder or flakes
- Infrared spectrum:Conforms
- Melting point:36 to 38 °C
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:2 g



