Pyrene 98%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:129-00-0
- Formula:C₁₆H₁₀
- MDL Number:MFCD00004136
- Molecular Weight:202.26 g/mol
- Merck Index:12,08147
- Boiling Pt:393 °C (760 mmHg)
- Melting Pt:148…152 °C
- Density:1,27 g/cm³ (20 °C)
- Flash Pt:210 °C
- Storage Temperature:Ambient
- EINECS:204-927-3
- UN:3077
- ADR:9,III
Dokumente
ACRO18083
SDS
Specifications
Specification Test Results
- Appearance:Light yellow to yellow to yellow-green Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:147 to 152 °C
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



