Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Boron trifluoride diethyl ether complex (∼48% BF₃)

Glass bottle for solids, 100 g
Catalog # ACRO174561000
Flammable
Corrosive
Irritant
Health
Boron trifluoride diethyl ether complex (∼48% BF₃)
Catalog # ACRO174561000
CAS Number:  109-63-7
Boron trifluoride diethyl ether complex (∼48% BF₃)
Boron trifluoride diethyl ether complex (∼48% BF₃)

Glass bottle for solids, 100 g

CAS Number:  109-63-7
Price
34,80 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Boron fluoride, ethyl ether complex, Boron trifluoride ethyl etherate, Boron trifluoride diethyl ether, Boron trifluoride etherate, Diethyl ether boron trifluoride, Boron trifluoride diethyl etherate
  • CAS Number:109-63-7
  • Formula:C₄H₁₀BF₃O
  • MDL Number:MFCD00013194
  • Molecular Weight:141.93 g/mol
  • Merck Index:13,01340
  • Boiling Pt:126 °C (760 mmHg)
  • Melting Pt:–60 °C
  • Density:1.12 g/cm³
  • Flash Pt:58 °C
  • Storage Temperature:Ambient
  • EINECS:203-689-8
  • UN:2604
  • ADR:8,I
Specifications

Specification Test Results

  • Appearance:
    Clear colorless to straw yellow Liquid (May darken on storage)
  • Refractive index:
    1.3429 to 1.3449 (20°C, 589 nm)
  • Assay:
    47.5 to 49 % (BF3)
  • GC:
    ≥97.5 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications