Boron trifluoride diethyl ether complex (∼48% BF₃)
Glass bottle, 2,5 kg
Boron trifluoride diethyl ether complex (∼48% BF₃)
ACRO174560025
: 109-63-7
: Boron fluoride, ethyl ether complex, Boron trifluoride ethyl etherate, Boron trifluoride diethyl ether, Boron trifluoride etherate, Diethyl ether boron trifluoride, Boron trifluoride diethyl etherate
- :109-63-7
- :C₄H₁₀BF₃O
- :MFCD00013194
- :141.93 g/mol
- :13,01340
- :126 °C (760 mmHg)
- :–60 °C
- :1.12 g/cm³
- :58 °C
- :Ambient
- :203-689-8
- :2604
- :8,I
- Appearance:Clear colorless to straw yellow Liquid (May darken on storage)
- Refractive index:1.3429 to 1.3449 (20°C, 589 nm)
- Assay:47.5 to 49 % (BF3)
- GC:≥97.5 %
- Pack type:Glass bottle
- Pk:2,5 kg



