2-Bromothiazole 98+%
Glass bottle, 25 mL
- :3034-53-5
- :C₃H₂BrNS
- :MFCD00005316
- :164.03 g/mol
- :70…71 °C (20 mmHg)
- :1.82 g/cm³
- :63 °C
- :Refrigerator
- :221-229-4
Documents
ACRO17091
SDS
- Appearance:Clear colorless to orange to light brown Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.5910 to 1.5930 (20°C, 589 nm)
- GC:≥98.0 %
- Pack type:Glass bottle
- Pk:25 mL
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



