2-Bromothiazole 98+%
Glass bottle, 5 mL
Supplier: Thermo Fisher Scientific
2-Bromothiazole 98+%
Catalog # ACRO170910050
Supplier: Thermo Fisher Scientific
CAS Number: 3034-53-5
Jump to:
Product Details & Documents
- CAS Number:3034-53-5
- Formula:C₃H₂BrNS
- MDL Number:MFCD00005316
- Molecular Weight:164.03 g/mol
- Boiling Pt:70…71 °C (20 mmHg)
- Density:1.82 g/cm³
- Flash Pt:63 °C
- Storage Temperature:Refrigerator
- EINECS:221-229-4
Specifications
Specification Test Results
- Appearance:Clear colorless to orange to light brown Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.5910 to 1.5930 (20°C, 589 nm)
- GC:≥98.0 %
Specifications
- Pack type:Glass bottle
- Pk:5 mL



