Order Entry
Germany
Orders LinkContactUsLinkComponent
L-Methionine 98+% | Avantor

L-Methionine 98+%

Plastic bottle, 100 g
Catalog # ACRO166161000
L-Methionine 98+%
Catalog # ACRO166161000
CAS Number:  63-68-3
L-Methionine 98+%
L-Methionine 98+%

Plastic bottle, 100 g

Wechseln zu:
CAS Number:  63-68-3
Price
88,80 €
Anmelden für die Verfügbarkeit
1 * 100 g
Product Details & Documents
Synonyme:  (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
  • CAS-Nummer:63-68-3
  • Formel::C₅H₁₁NO₂S
  • MDL-Nummer:MFCD00063097
  • Molekulargewicht:149.21 g/mol
  • Merck-Index:13,06004
  • Siedepunkt:278 °C (1013 hPa)
  • Schmelzpunkt:276…279 °C (dec.)
  • Dichte:1,373 g/cm³ (20 °C)
  • Lagertemperatur:Refrigerator
  • EINECS:200-562-9
Spezifikationen:

Spezifikation Testergebnisse

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.5 % (1 g, 105°C)
  • Sulfated ash:
    ≤0.4 %
  • TLC:
    ≤0.5 % of any individual impurity
  • pH:
    5.5 to 6.5 (1 % in water)
  • Specific optical rotation:
    +22.4° to +24.7° (20°C, 589 nm) (c=2, 6 N HCl)
  • Titration with HClO4:
    98.0 to 102.0 % (on dried basis)
  • Chloride (Cl):
    ≤500 ppm
  • Sulfate (SO4):
    ≤300 ppm
  • Iron (Fe):
    ≤30 ppm
  • Heavy metals (as Pb):
    ≤15 ppm

Spezifikationen:

  • Pack type:
    Plastic bottle
  • Pk:
    100 g

Spezifikationen: