Orders LinkContactUsLinkComponent
Stopcocks, straight through, with tip
Stopcocks, straight through, with tip
with polypropylene tip, 2 mm
Catalog # 661-1181
Supplier:   COWIE
Stopcocks, straight through, with tip
Catalog # 661-1181
Supplier:  COWIE
Stopcocks, straight through, with tip
Stopcocks, straight through, with tip
with polypropylene tip, 2 mm
Price
24,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
Made of PTFE.
  • Available with PP or glass tip
  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Hole Ø:
    2 mm
  • Description:
    with polypropylene tip