Ethyl crotonate 96%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: trans-Crotonic acid ethyl ester, 2-Butenoic acid ethyl ester, Crotonic acid ethyl ester, Ethyl trans-2-butenoate, trans-Ethyl crotonate, trans-2-Butenoic acid ethyl ester
- CAS Number:623-70-1
- Formula:C₆H₁₀O₂
- MDL Number:MFCD00009289
- Molecular Weight:114.14 g/mol
- Merck Index:13,02625
- Boiling Pt:143 °C (1013 hPa)
- Melting Pt:–79 °C
- Density:0.918 g/cm³
- Flash Pt:34 °C
- EINECS:210-808-7
- UN:1862
- ADR:3,II
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4220 to 1.4280 (20°C, 589 nm)
- Density:(20/4°C) 0.916 to 0.921 g/ml
- GC:≥95 %
- Acid value:≤1 mg KOH/g
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



