Order Entry
Germany
Orders LinkContactUsLinkComponent
Bromophenol blue, pure indicator

Bromophenol blue, pure indicator

Glass bottle for solids, 25 g
Catalog # ACRO151340250
Bromophenol blue, pure indicator
Catalog # ACRO151340250
CAS Number:  115-39-9
Bromophenol blue, pure indicator
Bromophenol blue, pure indicator

Glass bottle for solids, 25 g

CAS Number:  115-39-9
Price
94,40 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Merck Index:13,01429
  • Melting Pt:279 °C
  • Density:1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-086-2

Documents

Specifications

Specification Test Results

  • Appearance:
    Pink-orange to red Powder
  • Infrared spectrum:
    Conforms
  • Solubility:
    (0.1 % in ethanol) Clear
  • Specific extinction E (1%/1cm):
    ≥1100 (592 nm, in buffer)
  • Visual transition interval:
    from pH 3.0 (yellow) to pH 4.6 (blue)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.