Xanthine 98%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1H-Purine-2,6(3H,7H)-dione, 3,7-Dihydro-1H-purine-2,6-dione, 2,6-Dihydroxypurine
- CAS Number:69-89-6
- Formula:C₅H₄N₄O₂
- MDL Number:MFCD00078453
- Molecular Weight:152.11 g/mol
- Merck Index:12,10193
- Melting Pt:>300 °C
- Storage Temperature:Ambient
- EINECS:200-718-6
Documents
ACRO14917
SDS
Specifications
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 % (105°C, 5 hrs)
- Sulfated ash:≤0.2 % (800°C)
- HPLC:≥97.5 %
- TLC:Homogeneous
- UV spectrophotometry:98 to 102 % (lambda max. 278 nm, pH 10)
- Appearance of solution:(c=1, 1M NaOH) clear colorless to light yellow soln
- Heavy metals (as Pb):≤10 ppm
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



