Order Entry
Germany
Orders LinkContactUsLinkComponent
Uridine 99%

Uridine 99%

Glass bottle, 100 g
Catalog # ACRO140771000
Uridine 99%
Catalog # ACRO140771000
CAS Number:  58-96-8
Uridine 99%
Uridine 99%

Glass bottle, 100 g

CAS Number:  58-96-8
Price
556,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
  • CAS Number:58-96-8
  • Formula:C₉H₁₂N₂O₆
  • MDL Number:MFCD00006526
  • Molecular Weight:244.20 g/mol
  • Melting Pt:162…170 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-407-5

Documents

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    162 to 170 °C
  • Loss on drying:
    ≤1 % (105 °C, 2 hrs)
  • Sulfated ash:
    ≤0.1 %
  • HPLC:
    ≥98.5 %
  • UV spectrophotometry:
    ≥98 % (lambda max. 262 nm, pH 7)
  • UV transmittance:
    ≥95 % (c=1, H2O, 430 nm) 1 cm cell
  • Ratio of Absorbances:
    0.72 to 0.76 (A250/A260)
  • Heavy metals (as Pb):
    ≤10 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.