Lids for flat bottom chambers
Stainless steel lid for 200×200 mm and 200×100 mm twin trough and flat bottom chambers
Catalog # 552-0025
Supplier: CAMAG
Jump to:
Product Details & Documents
Specifications
- Description:Stainless steel lid for 200×200 mm and 200×100 mm twin trough and flat bottom chambers
- Seq:11