Order Entry
Germany
Orders LinkContactUsLinkComponent
D-(-)-Sorbitol 97%

D-(-)-Sorbitol 97%

Plastic bottle, 500 g
Catalog # ACRO132735000
D-(-)-Sorbitol 97%
Catalog # ACRO132735000
CAS Number:  50-70-4
D-(-)-Sorbitol 97%
D-(-)-Sorbitol 97%

Plastic bottle, 500 g

CAS Number:  50-70-4
Price
41,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004708
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,08797
  • Boiling Pt:295 °C (1013 hPa)
  • Melting Pt:95…99 °C
  • Density:1,524 g/cm³ (20 °C)
  • Flash Pt:≥ 100 °C
  • Storage Temperature:Ambient
  • EINECS:200-061-5

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤1.5 %
  • Sulfated ash:
    ≤0.1 %
  • HPLC:
    ≥96.0 % (on anhydrous substance)
  • Specific optical rotation:
    +4.0° to +7.0° (20°C, 589 nm) (c=10, borate soln.) (on anhydrous substance)
  • Lead (Pb):
    ≤0.5 ppm
  • Nickel (Ni):
    ≤1 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g

Specifications