Pyridine 99+%, extra pure
Glass bottle, 50 mL
Supplier: Thermo Fisher Scientific
Pyridine 99+%, extra pure
Catalog # ACRO131780500
Supplier: Thermo Fisher Scientific
CAS Number: 110-86-1
Jump to:
Product Details & Documents
Synonyms: Azabenzene, Azine
- CAS Number:110-86-1
- Formula:C₅H₅N
- MDL Number:MFCD00011732
- Molecular Weight:79.1 g/mol
- Merck Index:13,08060
- Boiling Pt:115…116 °C (760 mmHg)
- Melting Pt:–42 °C
- Density:0.978 g/cm³
- Flash Pt:17 °C
- Storage Temperature:Ambient
- EINECS:203-809-9
- UN:1282
- ADR:3,II
Documents
ACRO13178
SDS
ACRO131780250_TDS_EN
Product specification
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.5080 to 1.5110 (20°C, 589 nm)
- Water:≤0.1 % (Coulometric)
- GC:≥99.0 %
- Color scale:≤40 APHA
- Residue after evaporation:≤0.02 %
Specifications
- Pack type:Glass bottle
- Pk:50 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



