Order Entry
Germany
Orders LinkContactUsLinkComponent
Phthalide 99%

Phthalide 99%

Glass bottle for solids, 5 g
Catalog # ACRO131090050
Irritant
Phthalide 99%
Catalog # ACRO131090050
CAS Number:  87-41-2
Phthalide 99%
Phthalide 99%

Glass bottle for solids, 5 g

CAS Number:  87-41-2
Price
21,60 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Isobenzofuran-1(3H)-one
  • CAS Number:87-41-2
  • Formula:C₈H₆O₂
  • MDL Number:MFCD00005906
  • Molecular Weight:134.13 g/mol
  • Boiling Pt:290 °C (1013 hPa)
  • Melting Pt:70…73 °C
  • Density:1,1 g/cm³ (30 °C)
  • Flash Pt:152 °C
  • EINECS:201-744-0

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    70 to 73 °C
  • GC:
    ≥98.5 %
  • UV:
    at 230 nm A: ≤0.15
  • Color scale:
    ≤5 APHA
  • Residue after evaporation:
    ≤0.001 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.