Order Entry
Germany
Orders LinkContactUsLinkComponent
 

L-Phenylalanine 98.5+%

Plastic bottle, 25 g
  ACRO130310250
L-Phenylalanine 98.5+%
  ACRO130310250
 :  63-91-2
L-Phenylalanine 98.5+%
L-Phenylalanine 98.5+%

Plastic bottle, 25 g

 :  63-91-2
 
47,80 €
   
1 * 25 g
 
 :  H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine
  •  :63-91-2
  •  :C₉H₁₁NO₂
  •  :MFCD00064227
  •  :165.19 g/mol
  •  :14,07271
  •  :313 °C (1013 hPa)
  •  :270…275 °C (dec.)
  •  :0,754 g/cm³ (20 °C)
  •  :Ambient
  •  :200-568-1
 

 

  • Appearance:
    White to off-white Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.3 % (105°C, 3 hrs)
  • Sulfated ash:
    ≤0.1 %
  • pH:
    5.4 to 6.0 (1 % in water)
  • Specific optical rotation:
    -33.0° to -35.2° (20°C, 589 nm) (c=2, water) (on dried substance)
  • Titration with HClO4:
    98.50 to 102.00 % (On dried basis)
  • Lead (Pb):
    ≤10 ppm
  • Heavy metals (as Pb):
    ≤20 ppm

 

  • Pack type:
    Plastic bottle
  • Pk:
    25 g