Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Methylmorpholine 99%

4-Methylmorpholine 99%

Glass bottle for solids, 100 g
Catalog # ACRO127151000
Irritant
Corrosive
Flammable
4-Methylmorpholine 99%
Catalog # ACRO127151000
CAS Number:  109-02-4
4-Methylmorpholine 99%
4-Methylmorpholine 99%

Glass bottle for solids, 100 g

CAS Number:  109-02-4
Price
26,40 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  NMM, N-Methylmorpholine
  • CAS Number:109-02-4
  • Formula:C₅H₁₁NO
  • MDL Number:MFCD00006175
  • Molecular Weight:101.15 g/mol
  • Boiling Pt:115…116 °C (750 mmHg)
  • Melting Pt:–65 °C
  • Density:0.92 g/cm³
  • Flash Pt:14 °C
  • Storage Temperature:Ambient
  • EINECS:203-640-0
  • UN:2535
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4339 to 1.4359 (20°C, 589 nm)
  • Water:
    ≤0.5 % (K.F.)
  • GC:
    ≥98.5 %
  • Color scale:
    ≤50 APHA

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications