2'-Methoxyacetophenone 99%
Glass bottle, 25 mL
Supplier: Thermo Fisher Scientific
2'-Methoxyacetophenone 99%
Catalog # ACRO125670250
Supplier: Thermo Fisher Scientific
CAS Number: 579-74-8
Jump to:
Product Details & Documents
Synonyms: 1-(2-Methoxyphenyl)ethanone
- CAS Number:579-74-8
- Formula:C₉H₁₀O₂
- MDL Number:MFCD00008725
- Molecular Weight:150.18 g/mol
- Boiling Pt:131 °C (18 mmHg)
- Density:1.09 g/cm³
- Flash Pt:108 °C
- Storage Temperature:Ambient
- EINECS:209-446-2
Documents
ACRO12567
SDS
Specifications
Specification Test Results
- Appearance:Clear light yellow to orange to light brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5383 to 1.5403 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:25 mL



