4-Fluorophenol 99%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: p-Fluorophenol, 1-Fluoro-4-hydroxybenzene
- CAS Number:371-41-5
- Formula:C₆H₅FO
- MDL Number:MFCD00002316
- Molecular Weight:112.1 g/mol
- Boiling Pt:185 °C (1013 hPa)
- Melting Pt:44…49 °C
- Density:1,22 g/cm³ (20 °C)
- Flash Pt:68 °C
- Storage Temperature:Ambient
- EINECS:206-736-0
- UN:2811
- ADR:6.1,III
Specifications
Specification Test Results
- Appearance:Yellow to beige to pink Crystalline solid
- Infrared spectrum:Conforms
- Melting point:44 to 49 °C
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



