IL-1beta, Human, Biotrak Assay, ELISA - 96 wells (Range: 10-400pg/ml. Sensitivity: <1pg/ml. Sample volume: 50ul. 1.5L wash buffer su pplied. Calibrated to the WHO reference lot 86/552) 1 * 96 Assays
Catalog # RPN2751
Lieferant: Cytiva
Frequently Bought Together