Cyclopentane 97%, extra pure
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Cyclopentane 97%, extra pure
Catalog # ACRO111480010
Supplier: Thermo Fisher Scientific
CAS Number: 287-92-3
Jump to:
Product Details & Documents
Synonyms: Pentamethylene
- CAS Number:287-92-3
- Formula:C₅H₁₀
- MDL Number:MFCD00001356
- Molecular Weight:70.13 g/mol
- Merck Index:13,02769
- Boiling Pt:50 °C (760 mmHg)
- Melting Pt:–94 °C
- Density:0.75 g/cm³
- Flash Pt:–7 °C
- Storage Temperature:Ambient
- EINECS:206-016-6
- UN:1146
- ADR:3,II
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4050 to 1.4070 (20°C, 589 nm)
- GC:≥96.0 %
- Residue after evaporation:≤0.01 %
Specifications
- Pack type:Glass bottle
- Pk:1 L



