Cyclooctanone 98%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:502-49-8
- Formula:C₈H₁₄O
- MDL Number:MFCD00001754
- Molecular Weight:126.2 g/mol
- Boiling Pt:195…197 °C (760 mmHg)
- Melting Pt:39…44 °C
- Density:0.95 g/cm³
- Flash Pt:74 °C
- Storage Temperature:Ambient
- EINECS:207-940-2
- UN:1759
- ADR:8,III
Specifications
Specification Test Results
- Appearance:Colorless to white Low melting crystalline solid
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Titration with TBAH:≥97.5 %
- Color scale:≤50 APHA
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



