Anthrone ACS
Plastic bottle, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Anthracen-9(10H)-one
- CAS Number:90-44-8
- Formula:C₁₄H₁₀O
- MDL Number:MFCD00001187
- Molecular Weight:194.23 g/mol
- Boiling Pt:721 °C (760 mmHg)
- Melting Pt:152…157 °C
- Storage Temperature:Ambient
- EINECS:201-994-0
Documents
Specifications
Specification Test Results
- Appearance:Yellow Crystals
- Infrared spectrum:Conforms
- Melting point:152 to 157 °C
- Solubility:(in ethyl acetate) Passes test
- Absorbance:≤0.75 (425 nm, 1 cm cell)
- Sensitivity:(to carbohydrates) Passes test
Specifications
- Pack type:Plastic bottle
- Pk:25 g



