Orders LinkContactUsLinkComponent
Accessories
Accessories
Conductivity flow cell, titanium/EPDM/fluoro plastics, 14 μl volume, with M6 connection
Catalog # 18-1114-43
Supplier:   Cytiva
Accessories
Catalog # 18-1114-43
Supplier:  Cytiva
Accessories
Accessories
Conductivity flow cell, titanium/EPDM/fluoro plastics, 14 μl volume, with M6 connection
Price
979,00 €
Log in for availability
1 * 1 ST
Specifications
  • Description:
    Conductivity flow cell, titanium/EPDM/fluoro plastics, 14 μl volume, with M6 connection