Order Entry
Germany
Orders LinkContactUsLinkComponent
Containers with screw cap, multipurpose

Containers with screw cap, multipurpose

250 ml, 104 mm, 50 mm, Transparent
Catalog # BURK6202-0250
Supplier:   BURKLE
Containers with screw cap, multipurpose
Catalog # BURK6202-0250
Supplier:  BURKLE
Containers with screw cap, multipurpose
Containers with screw cap, multipurpose

250 ml, 104 mm, 50 mm, Transparent

Price
5,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP, translucent, with PE screw lid.

  • Suitable for use with foodstuffs
  • No crevices
  • Very good chemical and temperature resistance

Contents can be recognised easily due to the transparent sides. Emptying without residues.

Specifications
  • Ext.Ø:
    104 mm
  • Capacity:
    250 ml
  • Colour:
    Transparent
  • Height:
    50 mm