Orders LinkContactUsLinkComponent
Rotary evaporating flasks
Rotary evaporating flasks
2000 ml, 29/32 NS, non coated
Catalog # 531-1816
Supplier:   BUCHI
Rotary evaporating flasks
Catalog # 531-1816
Supplier:  BUCHI
Rotary evaporating flasks
Rotary evaporating flasks
2000 ml, 29/32 NS, non coated
Price
153,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Made of borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For Rotavapor® R-210/215 rotary evaporators. Pear-shaped with NS 29.2/32 ground joint socket.
Specifications
  • Socket size:
    29/32 NS
  • Type:
    non coated
  • Capacity:
    2000 ml
  • Seq:
    58