Rotary evaporating flasks
50 ml, 29/32 NS, non coated
Catalog # 531-1811
Supplier: BUCHI
Jump to:
Product Details & Documents
Made of borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For Rotavapor® R-210/215 rotary evaporators. Pear-shaped with NS 29.2/32 ground joint socket.
Specifications
- Socket size:29/32 NS
- Type:non coated
- Capacity:50 ml
- Seq:50